Merck
  • A novel strategy for dual-channel detection of metallothioneins and mercury based on the conformational switching of functional chimera aptamer.
A novel strategy for dual-channel detection of metallothioneins and mercury based on the conformational switching of functional chimera aptamer.
Journal of pharmaceutical and biomedical analysis (2015-01-28)
Xian Tang, Yong-Sheng Wang, Jin-Hua Xue, Bin Zhou, Jin-Xiu Cao, Si-Han Chen, Ming-Hui Li, Xiao-Feng Wang, Yu-Feng Zhu, Yan-Qin Huang
ZUSAMMENFASSUNG
A novel strategy for dual-channel detection of metallothioneins (MTs) and Hg(2+) has been proposed. In the absence of Hg(2+), the functional chimera aptamer (FCA) designed can form an intact G-quadruplex with flexibility, which was demonstrated to have peroxidase-like activities upon hemin binding. In the presence of Hg(2+), the formation of T-Hg(2+)-T complex results in the conformational switching of FCA, which lost the peroxidase-like activities and cannot catalyze the oxidation of ABTS by H2O2. Upon addition of MTs in this solution, MTs could interact with Hg(2+) to form a MTs-Hg(2+) complex, leading to the recovery of the G-quadruplex DNAzyme. The color and absorbance of the sensing system were also changed accordingly. In the optimizing condition, ΔA was directly proportional to the concentration ranging from 8.84 nM to 1.0 μM for Hg(2+), and 7.82 nM to 0.462 μM for MTs with the detection limits of 2.65 nM and 2.34 nM, respectively. The proposed dual-channel method avoids the label steps in common methods, and allows direct analysis of the samples without costly instruments, and is reliable, inexpensive and sensitive.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumnitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Hämin, from bovine, ≥90%
Sigma-Aldrich
Hämin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Kaliumnitrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Kaliumnitrat, 99.999% trace metals basis
Sigma-Aldrich
Kaliumnitrat, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, 99.99% trace metals basis
Supelco
Mettler-Toledo-Kalibriersubstanz ME 51143095, Kaliumnitrat, traceable to primary standards (LGC)