Merck
  • Plasma cardiac troponin I concentration and cardiac death in cats with hypertrophic cardiomyopathy.
Plasma cardiac troponin I concentration and cardiac death in cats with hypertrophic cardiomyopathy.
Journal of veterinary internal medicine (2014-10-17)
K Borgeat, K Sherwood, J R Payne, V Luis Fuentes, D J Connolly
ZUSAMMENFASSUNG
The use of cardiac biomarkers to assist in the diagnosis of occult and symptomatic hypertrophic cardiomyopathy (HCM) in cats has been established. There is limited data describing their prognostic utility in cats with HCM. Circulating concentrations of N-terminal B-type natriuretic peptide (NTproBNP) and cardiac troponin I (cTnI) predict cardiac death in cats with HCM. Forty-one cats diagnosed with HCM at a veterinary teaching hospital, between February 2010 and May 2011. Prospective investigational study. Plasma samples were collected from cats diagnosed with HCM and concentrations of NTproBNP and cTnI were analyzed at a commercial laboratory. Echocardiographic measurements from the day of blood sampling were recorded. Long-term outcome data were obtained. Associations with time to cardiac death were analyzed using Cox proportional hazards models. When controlling for the presence/absence of heart failure and echocardiographic measures of left atrial size and function, cTnI > 0.7 ng/mL was independently associated with time to cardiac death. In univariable analysis, NTproBNP > 250 pmol/L was associated with cardiac death (P = .023), but this did not remain significant (P = .951) when controlling for the effect of clinical signs or left atrial size/function. Plasma concentration of cTnI (cutoff >0.7 ng/mL) is a predictor of cardiac death in cats with HCM that is independent of the presence of heart failure or left atrial dilatation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Levothyroxin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Thyroxin Natriumsalz Pentahydrat, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxin Natriumsalz Pentahydrat, γ-irradiated, powder, BioXtra, suitable for cell culture