Merck
Antiviral effect of cidofovir on parvovirus B19 replication.
Antiviral research (2014-12-03)
Francesca Bonvicini, Gloria Bua, Elisabetta Manaresi, Giorgio Gallinella
ZUSAMMENFASSUNG
Parvovirus B19 (B19V) is a human ssDNA virus responsible for a wide range of clinical manifestations, still lacking for a specific antiviral therapy. The identification of compounds active against B19V may add therapeutic options to the treatment of B19V infections, that now entirely relies on symptomatic treatments. In the search for compounds possibly inhibiting B19V replication, a particular focus was raised to cidofovir, an acyclic nucleoside phosphonate broadly active against dsDNA viruses. The present study was aimed at evaluating the effect of cidofovir against B19V in two model systems, the UT7/EpoS1 cell line and erythroid progenitor cells (EPC), generated from peripheral blood mononuclear cells. Experiments were carried out at different multiplicity of infections and cidofovir concentrations (0-500 μM) during a course of infection. The effects of cidofovir on B19V replication were assessed by qPCR assays while influence of cidofovir on host cells was measured by cell proliferation and viability assays. Our findings demonstrated that cidofovir has a relevant inhibiting activity on B19V replication within infected UT7/EpoS1, and that the effect on B19V DNA amounts is dose-dependent allowing for the determination of EC50 and EC90 values (7.45-41.27 μM, and 84.73-360.7 μM, respectively). In EPCs, that constitute a cellular population close to the natural target cells in bone marrow, the inhibitory effect was demonstrated to a lesser extent, however provoking a significant reduction on B19V DNA amounts at 500 μM (68.2-92.8%). To test infectivity of virus released from EPCs cultured in the presence of cidofovir, cell culture supernatants were used as inoculum for a further course of infection in UT7/EpoS1 cells, indicating a significant reduction in viral infectivity at 500 μM cidofovir. Since the drug did not interfere with the overall cellular DNA synthesis and metabolic activity, the observed effect of cidofovir could be likely related to a specific inhibition of B19V replication.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Aceton, analytical standard
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-Brom-2′-deoxyuridin, BioUltra, ≥99%
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Anti-Parvovirus B19-Antikörper, A.S. 328-344 von VP2-Kapsidprotein, Klon R92F6, clone R92F6, Chemicon®, from mouse
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.