Merck
  • In-vitro maturation versus IVF with GnRH antagonist for women with polycystic ovary syndrome: treatment outcome and rates of ovarian hyperstimulation syndrome.
In-vitro maturation versus IVF with GnRH antagonist for women with polycystic ovary syndrome: treatment outcome and rates of ovarian hyperstimulation syndrome.
Reproductive biomedicine online (2014-09-30)
Mausumi Das, Weon-Young Son, William Buckett, Togas Tulandi, Hananel Holzer
ZUSAMMENFASSUNG
In-vitro maturation (IVM) treatment has gained popularity for decreasing the incidence of ovarian hyperstimulation syndrome (OHSS) by eliminating or minimizing the use of gonadotrophins in women with polycystic ovary syndrome (PCOS). Studies have shown that IVF with GnRH-antagonist protocol is associated with a lower incidence of OHSS. Data comparing the relative success of these two treatments is, however, lacking. Treatment outcome and rates of OHSS were compared in patients with PCOS who underwent assisted conception with either IVM or IVF with GnRH-antagonist protocol between 2006 and 2011. The number of oocytes retrieved was higher in the IVM group, whereas the number of mature oocytes, fertilization rate and number of embryos cleaved were comparable. The implantation rate was higher in the IVF group. The clinical pregnancy rates per embryo transfer were not statistically different (IVF: 45.8% versus IVM: 32.4%). The live-birth rate was higher in the IVF group (IVF: 40.7% versus IVM: 23.5%; P = 0.04). Five women developed moderate or severe OHSS in the IVF group, whereas none did in the IVM group. Both IVM and IVF with GnRH-antagonist protocol seem to be effective treatment regimens in women with PCOS, although IVM is associated with a lower risk of OHSS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
Progesteron, meets USP testing specifications
Sigma-Aldrich
Progesteron, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Progesteron, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Progesteron, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Luteinisierendes Hormon Releasing-Hormon (LHRH), ≥97% (HPLC)
Sigma-Aldrich
Luteinizing hormone releasing hormone salmon, ≥97% (HPLC)
Progesteron für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Progesteron, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesteron, VETRANAL®, analytical standard
Progesteron für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard