Merck
Cytotoxic saponins from the seeds of Pittosporum angustifolium.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2014-07-30)
Christian Bäcker, Kristina Jenett-Siems, Karsten Siems, Martina Wurster, Anja Bodtke, Ulrike Lindequist
ZUSAMMENFASSUNG
Three new acylated R1-barrigenol triterpene glycosides, 1-3, were isolated from the seeds of Pittosporum angustifolium Lodd. together with four known glycosides, 4-7, containing R1- and A1-barrigenol backbones. On the basis of spectroscopic, spectrometric, and chemical analyses the novel compounds were named pittangretosides N-P and established as 21beta-acetoxy-22alpha-angeloyloxy- (1), 21beta-acetoxy-22alpha-(2-acetoxy-2-methylbutyroyloxy)- (2), and 21beta-(2-methylbutyroyloxy)-22alpha-acetoxy-3beta-[beta-D-glucopyranosyl- (1 --> 2)]-[alpha-L-arabinopyranosyl-(1 --> 3)]-[alpha-L-arabinofuranosyl-(1 --> 4)]-beta-D-glucuronopyranosyloxyolean-12-ene-15alpha, 6alpha, 28-triol (3). Evaluation of the in vitro cytotoxicity against three tumour cell lines and one non-tumourigenic cell line revealed antiproliferative effects with IC50 values in a range of 1.74-34.1 microM.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
D-Glucuronsäure, ≥98% (GC)
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Thymol, ≥98.5%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
Etoposid, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Thymol, FCC, FG
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Supelco
Thymol, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Supelco
Thymol, analytical standard
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Galactose, European Pharmacopoeia (EP) Reference Standard
Supelco
Thymol, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Etoposid, European Pharmacopoeia (EP) Reference Standard
Thymol, European Pharmacopoeia (EP) Reference Standard
Etoposid für die Systemeignung, European Pharmacopoeia (EP) Reference Standard