Merck
Succinate causes pathological cardiomyocyte hypertrophy through GPR91 activation.
Cell communication and signaling : CCS (2014-12-30)
Carla J Aguiar, João A Rocha-Franco, Pedro A Sousa, Anderson K Santos, Marina Ladeira, Cibele Rocha-Resende, Luiz O Ladeira, Rodrigo R Resende, Fernando A Botoni, Marcos Barrouin Melo, Cristiano X Lima, José M Carballido, Thiago M Cunha, Gustavo B Menezes, Silvia Guatimosim, M Fatima Leite
ZUSAMMENFASSUNG
Succinate is an intermediate of the citric acid cycle as well as an extracellular circulating molecule, whose receptor, G protein-coupled receptor-91 (GPR91), was recently identified and characterized in several tissues, including heart. Because some pathological conditions such as ischemia increase succinate blood levels, we investigated the role of this metabolite during a heart ischemic event, using human and rodent models. We found that succinate causes cardiac hypertrophy in a GPR91 dependent manner. GPR91 activation triggers the phosphorylation of extracellular signal-regulated kinase 1/2 (ERK1/2), the expression of calcium/calmodulin dependent protein kinase IIδ (CaMKIIδ) and the translocation of histone deacetylase 5 (HDAC5) into the cytoplasm, which are hypertrophic-signaling events. Furthermore, we found that serum levels of succinate are increased in patients with cardiac hypertrophy associated with acute and chronic ischemic diseases. These results show for the first time that succinate plays an important role in cardiomyocyte hypertrophy through GPR91 activation, and extend our understanding of how ischemia can induce hypertrophic cardiomyopathy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Bernsteinsäure, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Bernsteinsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, ReagentPlus®, BioRenewable, ≥99.0%
Supelco
Bernsteinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bernsteinsäure, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Bernsteinsäure, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
KN-93, ≥98% (HPLC)
Supelco
Bernsteinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland