Merck
  • Role of protein phosphatase magnesium-dependent 1A and anti-protein phosphatase magnesium-dependent 1A autoantibodies in ankylosing spondylitis.
Role of protein phosphatase magnesium-dependent 1A and anti-protein phosphatase magnesium-dependent 1A autoantibodies in ankylosing spondylitis.
Arthritis & rheumatology (Hoboken, N.J.) (2014-07-02)
Yong-Gil Kim, Dong Hyun Sohn, Xiaoyan Zhao, Jeremy Sokolove, Tamsin M Lindstrom, Bin Yoo, Chang-Keun Lee, John D Reveille, Joel D Taurog, William H Robinson
ZUSAMMENFASSUNG
Although ankylosing spondylitis (AS) is driven by immune-mediated processes, little is known about the presence and role of autoantibodies in this disease. This study was undertaken to investigate whether autoantibodies occur in and are involved in AS. We performed human protein microarray analysis of sera derived from patients with AS or other autoimmune disorders to identify autoantibodies associated specifically with AS, and identified autoantibody targeting of protein phosphatase magnesium-dependent 1A (PPM1A) in AS. We performed enzyme-linked immunosorbent assay (ELISA) analysis of sera from 2 independent AS cohorts to confirm autoantibody targeting of PPM1A, and to assess associations between levels of anti-PPM1A antibodies and AS disease severity or response to anti-tumor necrosis factor (anti-TNF) therapy (as measured by Bath AS Disease Activity Index [BASDAI] score). Levels of anti-PPM1A antibodies were also evaluated in sera from rats transgenic for HLA-B27 and human β2 -microglobulin. The expression of PPM1A was assessed by immunohistochemistry in synovial tissue samples from patients with AS, rheumatoid arthritis, or osteoarthritis. The role of PPM1A in osteoblast differentiation was investigated by gene knockdown and overexpression. AS was associated with autoantibody targeting of PPM1A, and levels of anti-PPM1A autoantibodies were significantly higher in patients with more advanced sacroiliitis and correlated positively with BASDAI score after treatment with anti-TNF agents. The levels of anti-PPM1A autoantibodies were also higher in the sera of transgenic rats that are prone to develop spondyloarthritis than in those that are not. PPM1A was expressed in AS synovial tissue, and PPM1A overexpression promoted osteoblast differentiation, whereas PPM1A knockdown suppressed it. Anti-PPM1A autoantibodies are present in AS, and our findings suggest that PPM1A may contribute to the pathogenic bone ankylosis characteristic of AS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
SAFC
BIS-TRIS
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Ascorbinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbinsäure, tested according to Ph. Eur.