Merck
  • Elasticity of nanoparticles influences their blood circulation, phagocytosis, endocytosis, and targeting.
Elasticity of nanoparticles influences their blood circulation, phagocytosis, endocytosis, and targeting.
ACS nano (2015-02-27)
Aaron C Anselmo, Mengwen Zhang, Sunny Kumar, Douglas R Vogus, Stefano Menegatti, Matthew E Helgeson, Samir Mitragotri
ZUSAMMENFASSUNG
The impact of physical and chemical modifications of nanoparticles on their biological function has been systemically investigated and exploited to improve their circulation and targeting. However, the impact of nanoparticles' flexibility (i.e., elastic modulus) on their function has been explored to a far lesser extent, and the potential benefits of tuning nanoparticle elasticity are not clear. Here, we describe a method to synthesize polyethylene glycol (PEG)-based hydrogel nanoparticles of uniform size (200 nm) with elastic moduli ranging from 0.255 to 3000 kPa. These particles are used to investigate the role of particle elasticity on key functions including blood circulation time, biodistribution, antibody-mediated targeting, endocytosis, and phagocytosis. Our results demonstrate that softer nanoparticles (10 kPa) offer enhanced circulation and subsequently enhanced targeting compared to harder nanoparticles (3000 kPa) in vivo. Furthermore, in vitro experiments show that softer nanoparticles exhibit significantly reduced cellular uptake in immune cells (J774 macrophages), endothelial cells (bEnd.3), and cancer cells (4T1). Tuning nanoparticle elasticity potentially offers a method to improve the biological fate of nanoparticles by offering enhanced circulation, reduced immune system uptake, and improved targeting.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cyclohexan, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexan, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Rhodamin B, ≥95% (HPLC)
Sigma-Aldrich
2-Hydroxy-2-methylpropiophenon, 97%
Sigma-Aldrich
Rhodamin B, suitable for fluorescence
Sigma-Aldrich
2-Carboxyethylacrylat, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Cyclohexan, anhydrous, 99.5%
Sigma-Aldrich
Cyclohexan, ACS reagent, ≥99%
Supelco
Cyclohexan, analytical standard
Supelco
Rhodamin B, analytical standard
Supelco
Rhodamin B -Lösung, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Rhodamin B, Dye content 90 %