Merck
REDD1 attenuates cardiac hypertrophy via enhancing autophagy.
Biochemical and biophysical research communications (2014-12-03)
Chen Liu, Ruicong Xue, Dexi Wu, Lingling Wu, Cong Chen, Weiping Tan, Yili Chen, Yugang Dong
ZUSAMMENFASSUNG
Cardiac hypertrophy is a major risk factor of cardiovascular morbidity and mortality. Autophagy is established to be involved in regulating cardiac hypertrophy. REDD1, a stress-responsive protein, is proved to contribute in autophagy induction. However, the role of REDD1 in cardiac hypertrophy remains unknown. Our study demonstrated that REDD1 knockdown by RNAi exacerbated phenylephrine (PE)-induced cardiac hypertrophy, manifested by increased hypertrophic markers such as ANP and cell surface area. In addition, we discovered that ERK1/2 signaling pathway was involved in the effect of REDD1 on hypertrophy. Moreover, our study showed that REDD1 knockdown impaired autophagy in hypertrophied cardiomyocytes. mTOR, a signaling molecule governing autophagy induction, was activated by the knockdown of REDD1 under PE stress. Importantly, the pro-hypertrophic effect of REDD1 knockdown was significantly reversed by the autophagy enhancer rapamycin. Taken together, we firstly prove that REDD1 is essential for inhibiting cardiac hypertrophy by enhancing autophagy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Supelco
Rapamycin, VETRANAL®, analytical standard