Merck
  • Designing versatile heterogeneous catalysts based on Ag and Au nanoparticles decorated on chitosan functionalized graphene oxide.
Designing versatile heterogeneous catalysts based on Ag and Au nanoparticles decorated on chitosan functionalized graphene oxide.
Physical chemistry chemical physics : PCCP (2015-04-08)
Rajendiran Rajesh, E Sujanthi, S Senthil Kumar, Rengarajan Venkatesan
ZUSAMMENFASSUNG
Herein we report the covalent grafting of chitosan on graphene oxide (GO) followed by a simple approach for anchoring silver (AgNPs) and gold (AuNPs) nanoparticles onto a chitosan grafted graphene oxide surface by a NaBH4 reduction method. Catalytic activity of prepared heterogeneous GO grafted chitosan stabilized silver and gold nanocatalysts (GO-Chit-Ag/AuNPs) was explored for the reduction of aromatic nitroarenes and degradation of hazardous azo dyes in the presence of NaBH4. Both catalysts were found to exhibit excellent catalytic activity towards the reduction of aromatic nitroarenes and azo dyes degradation. Furthermore, the nanocatalysts were found to be selective towards the reduction of nitro groups in halonitroarenes without any dehalogenation under mild conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Natriumborhydrid, powder, ≥98.0%
Sigma-Aldrich
Natriumnitrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumnitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Natriumborhydrid, ReagentPlus®, 99%
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Thionylchlorid, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumborhydrid, granular, 99.99% trace metals basis
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Methylorange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Natriumborhydrid, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Thionylchlorid, reagent grade, 97%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Methylorange, suitable for microscopy
Sigma-Aldrich
Natriumnitrat, 99.995% trace metals basis
Sigma-Aldrich
Natriumborhydrid, granular, 10-40 mesh, 98%
Sigma-Aldrich
Natriumborhydrid -Lösung, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Natriumnitrat, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Natriumborhydrid, caplets (18 × 10 × 8 mm), 98%
Supelco
Natriumnitrat, analytical standard
Sigma-Aldrich
Natriumborhydrid -Lösung, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Natriumnitrat, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, tested according to Ph. Eur.
Sigma-Aldrich
Natriumnitrat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumnitrat-14N, 99.95 atom % 14N