Merck
A rapid method for obtaining mesenchymal stem cells and platelets from bone marrow aspirate.
Journal of tissue engineering and regenerative medicine (2012-06-21)
Barbara Dozza, Giuliana Gobbi, Enrico Lucarelli, Michela Pierini, Claudia Di Bella, Tommaso Frisoni, Pier Luigi Tazzari, Francesca Ricci, Prisco Mirandola, Cecilia Carubbi, Sandro Giannini, Davide Donati, Marco Vitale
ZUSAMMENFASSUNG
Mesenchymal stem cells (MSCs) and platelet-rich plasma (PRP) are currently used alone or in combination for therapeutic applications especially for bone repair. We tested whether MSCs can be isolated from bone marrow (BM) aspirate using a commercially available kit commonly used to obtain PRP from peripheral blood (PB). Results revealed that mononuclear cells and platelets from both PB and BM could be efficiently isolated by obtaining a mononuclear and platelet rich fraction (PB-MPRF and BM-MPRF, respectively). Starting with comparable volumes, the number of platelets increased 1.5-fold in BM-MPRF compared to PB-MPRF. The number of clonogenic cells in BM-MPRF samples was significantly higher than whole BM samples as revealed by CFU-F assay (54.92 ± 8.55 CFU-F/1.5 x 10(5) nucleated cells and 32.50 ± 12.43 CFU-F/1.5 x 10(5) nucleated cells, respectively). Cells isolated from BM-MPRF after in vitro expansion fulfilled the definition of MSCs by phenotypic criteria, and differentiated along osteogenic, adipogenic and chondrogenic lineages following induction. Results showed that the kit isolated MSCs and platelets from BM aspirate. Isolated MSCs were further expanded in a laboratory and BM-MPRF was used clinically following BM withdrawal for rapid intra-operative cell therapy for the treatment of bone defects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, ≥99% (HPLC), powder
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, BioUltra, ≥99%
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Fluorescein, free acid
Supelco
Natriumzitrat, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cetylpyridiniumchlorid, meets USP testing specifications
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Dexamethason, meets USP testing specifications
USP
Cetylpyridiniumchlorid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioUltra, Molecular Biology, 1 M in H2O
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Cetylpyridiniumchlorid, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethason, tested according to Ph. Eur.
Dexamethason, British Pharmacopoeia (BP) Assay Standard
Dexamethason für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Dexamethason für die Systemeignung, European Pharmacopoeia (EP) Reference Standard