Merck
  • Plasmid-encoded Pgp3 is a major virulence factor for Chlamydia muridarum to induce hydrosalpinx in mice.
Plasmid-encoded Pgp3 is a major virulence factor for Chlamydia muridarum to induce hydrosalpinx in mice.
Infection and immunity (2014-10-08)
Yuanjun Liu, Yumeng Huang, Zhangsheng Yang, Yina Sun, Siqi Gong, Shuping Hou, Chaoqun Chen, Zhongyu Li, Quanzhong Liu, Yimou Wu, Joel Baseman, Guangming Zhong
ZUSAMMENFASSUNG
Hydrosalpinx induction in mice by Chlamydia muridarum infection, a model that has been used to study C. trachomatis pathogenesis in women, is known to depend on the cryptic plasmid that encodes eight genes designated pgp1 to pgp8. To identify the plasmid-encoded pathogenic determinants, we evaluated C. muridarum transformants deficient in the plasmid-borne gene pgp3, -4, or -7 for induction of hydrosalpinx. C. muridarum transformants with an in-frame deletion of either pgp3 or -4 but not -7 failed to induce hydrosalpinx. The deletion mutant phenotype was reproduced by using transformants with premature termination codon insertions in the corresponding pgp genes (to minimize polar effects inherent in the deletion mutants). Pgp4 is known to regulate pgp3 expression, while lack of Pgp3 does not significantly affect Pgp4 function. Thus, we conclude that Pgp3 is an effector virulence factor and that lack of Pgp3 may be responsible for the attenuation in C. muridarum pathogenicity described above. This attenuated pathogenicity was further correlated with a rapid decrease in chlamydial survival in the lower genital tract and reduced ascension to the upper genital tract in mice infected with C. muridarum deficient in Pgp3 but not Pgp7. The Pgp3-deficient C. muridarum organisms were also less invasive when delivered directly to the oviduct on day 7 after inoculation. These observations demonstrate that plasmid-encoded Pgp3 is required for C. muridarum survival in the mouse genital tract and represents a major virulence factor in C. muridarum pathogenesis in mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Glutaminsäure, European Pharmacopoeia (EP) Reference Standard