Merck
  • A cholinergic contribution to the circulatory responses evoked at the onset of handgrip exercise in humans.
A cholinergic contribution to the circulatory responses evoked at the onset of handgrip exercise in humans.
American journal of physiology. Regulatory, integrative and comparative physiology (2015-01-16)
Lauro C Vianna, Paul J Fadel, Niels H Secher, James P Fisher
ZUSAMMENFASSUNG
A cholinergic (muscarinic) contribution to the initial circulatory response to exercise in humans remains controversial. Herein, we posit that this may be due to exercise mode with a cholinergic contribution being important during isometric handgrip exercise, where the hyperemic response of the muscle is relatively small compared with the onset of leg cycling, where a marked increase in muscle blood flow rapidly occurs as a consequence of multiple redundant mechanisms. We recorded blood pressure (BP; brachial artery), stroke volume (pulse contour analysis), cardiac output, and systemic vascular resistance (SVR) in young healthy males, while performing either 20 s of isometric handgrip contraction at 40% maximum voluntary contraction (protocol 1; n = 9) or 20 s of low-intensity leg cycling exercise (protocol 2; n = 8, 42 ± 8 W). Exercise trials were conducted under control (no drug) conditions and following cholinergic blockade (glycopyrrolate). Under control conditions, isometric handgrip elicited an initial increase in BP (+5 ± 2 mmHg at 3 s and +3 ± 1 mmHg at 10 s, P < 0.05), while SVR dropped after 3 s (-27 ± 6% at 20 s; P < 0.05). Cholinergic blockade abolished the isometric handgrip-induced fall in SVR and, thereby, augmented the pressor response (+13 ± 3 mmHg at 10 s; P < 0.05 vs. control). In contrast, cholinergic blockade had a nonsignificant effect on changes in BP and SVR at the onset of leg cycling exercise. These findings suggest that a cholinergic mechanism is important for the BP and SVR responses at the onset of isometric handgrip exercise in humans.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lidocain, powder
Supelco
Lidocain, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lidocain, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lidocain, analytical standard
Sigma-Aldrich
Glycopyrroniumbromid, ≥98% (HPLC)
Lidocain, European Pharmacopoeia (EP) Reference Standard
Glycopyrroniumbromid, European Pharmacopoeia (EP) Reference Standard
Glycopyrronium für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Glycopyrronium Unreinheit N, European Pharmacopoeia (EP) Reference Standard