Merck
  • Evaluation of levobupivacaine passage to breast milk following epidural anesthesia for cesarean delivery.
Evaluation of levobupivacaine passage to breast milk following epidural anesthesia for cesarean delivery.
International journal of obstetric anesthesia (2014-06-24)
E Bolat, A Bestas, M K Bayar, S Ozcan, O L Erhan, B Ustundag
ZUSAMMENFASSUNG
Following maternal administration, local anesthetics pass into breast milk. In the present study, we aimed to compare the passage of levobupivacaine and bupivacaine into breast milk following epidural anesthesia for cesarean delivery. A total of 20 women undergoing elective cesarean delivery under epidural anesthesia were randomized to receive either 0.5% levobupivacaine or 0.5% racemic bupivacaine via an epidural catheter. Immediately before and 30min, 1h, 2h, 6h, 12h and 24h after administration of epidural local anesthetic, maternal blood and breast milk samples were taken simultaneously. Drug concentrations in plasma and milk were determined via high-performance liquid chromatography. The infant's drug exposure was determined by calculating milk/plasma ratios of levobupivacaine and bupivacaine. Both levobupivacaine and bupivacaine were detected in breast milk 30min after epidural administration. Concentrations of both agents showed constant and similar decreases in milk and plasma and were nearly undetectable at 24h. The milk/plasma ratios were 0.34±0.13 for levobupivacaine and 0.37±0.14 for bupivacaine. Both levobupivacaine and bupivacaine pass into breast milk following epidural administration. The concentration of both drugs was approximately three times lower in breast milk than in maternal plasma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lidocain, powder
Supelco
Lidocain, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Lidocain, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lidocain, analytical standard
Lidocain, European Pharmacopoeia (EP) Reference Standard