Merck
  • Mitochondrial Ca2+-dependent NLRP3 activation exacerbates the Pseudomonas aeruginosa-driven inflammatory response in cystic fibrosis.
Mitochondrial Ca2+-dependent NLRP3 activation exacerbates the Pseudomonas aeruginosa-driven inflammatory response in cystic fibrosis.
Nature communications (2015-02-05)
Alessandro Rimessi, Valentino Bezzerri, Simone Patergnani, Saverio Marchi, Giulio Cabrini, Paolo Pinton
ZUSAMMENFASSUNG
The common pathological manifestation of cystic fibrosis (CF) is associated with an excessive lung inflammatory response characterized by interleukin-1β accumulation. CF airway epithelial cells show an exacerbated pro-inflammatory response to Pseudomonas aeruginosa; however, it is unclear whether this heightened inflammatory response is intrinsic to cells lacking CF transmembrane conductance regulator (CFTR). Here we demonstrate that the degree and quality of the inflammatory response in CF are supported by P. aeruginosa-dependent mitochondrial perturbation, in which flagellin is the inducer and mitochondrial Ca(2+) uniporter (MCU) is a signal-integrating organelle member for NLRP3 activation and IL-1β and IL-18 processing. Our work elucidates the regulation of the NLRP3 inflammasome by mitochondrial Ca(2+) in the P. aeruginosa-dependent inflammatory response and deepens our understanding of the significance of mitochondria in the Ca(2+)-dependent control of inflammation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumfluorid, ACS reagent, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Natriumfluorid, ReagentPlus®, ≥99%
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumfluorid, 99.99% trace metals basis
Supelco
Natriumfluorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumfluorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Natriumfluorid, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Mechlorethamin -hydrochlorid, 98%
Sigma-Aldrich
Natriumfluorid, BioXtra, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Natriumfluoridlösung
Sigma-Aldrich
Natriumfluorid, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumfluorid 0,5 M -Lösung
Supelco
Fluorid-Ionenlösung für ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Tetramethylrhodamin-methylester-perchlorat, ≥95%
Natriumlaurylsulfat, European Pharmacopoeia (EP) Reference Standard