Merck
  • Natural genetic variation in Arabidopsis thaliana defense metabolism genes modulates field fitness.
Natural genetic variation in Arabidopsis thaliana defense metabolism genes modulates field fitness.
eLife (2015-04-14)
Rachel Kerwin, Julie Feusier, Jason Corwin, Matthew Rubin, Catherine Lin, Alise Muok, Brandon Larson, Baohua Li, Bindu Joseph, Marta Francisco, Daniel Copeland, Cynthia Weinig, Daniel J Kliebenstein
ZUSAMMENFASSUNG
Natural populations persist in complex environments, where biotic stressors, such as pathogen and insect communities, fluctuate temporally and spatially. These shifting biotic pressures generate heterogeneous selective forces that can maintain standing natural variation within a species. To directly test if genes containing causal variation for the Arabidopsis thaliana defensive compounds, glucosinolates (GSL) control field fitness and are therefore subject to natural selection, we conducted a multi-year field trial using lines that vary in only specific causal genes. Interestingly, we found that variation in these naturally polymorphic GSL genes affected fitness in each of our environments but the pattern fluctuated such that highly fit genotypes in one trial displayed lower fitness in another and that no GSL genotype or genotypes consistently out-performed the others. This was true both across locations and within the same location across years. These results indicate that environmental heterogeneity may contribute to the maintenance of GSL variation observed within Arabidopsis thaliana.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-(Diethylamino)ethanol, ≥99.5%