Merck
  • Tear exchangeable limbal rigid contact lens for ocular sequelae resulting from Stevens-Johnson syndrome or toxic epidermal necrolysis.
Tear exchangeable limbal rigid contact lens for ocular sequelae resulting from Stevens-Johnson syndrome or toxic epidermal necrolysis.
American journal of ophthalmology (2014-07-19)
Chie Sotozono, Naoki Yamauchi, Soshun Maeda, Shigeru Kinoshita
ZUSAMMENFASSUNG
To evaluate the therapeutic benefits of tear-exchangeable, limbal, rigid contact lenses (limbal CLs) in patients with Stevens-Johnson syndrome- or toxic epidermal necrolysis-associated ocular sequelae. Noncomparative, retrospective, interventional case series. We enrolled 53 eyes of 42 patients (mean age, 51.8 ± 13.9 years; mean follow-up, 25.7 ± 15.7 months) with Stevens-Johnson syndrome- or toxic epidermal necrolysis-associated ocular sequelae and divided them into 3 groups according to the best-corrected visual acuity (BCVA) before limbal CL fitting: (1) BCVA worse than 20/2000 (11 eyes), (2) BCVA ranging from 20/200 to 20/2000 (31 eyes), and (3) BCVA of 20/200 or better (11 eyes). The BCVA and the 25-item National Eye Institute Visual Function Questionnaire (NEI VFQ-25) composite score before fitting and after 3 months of limbal CL use were evaluated. The change in BCVA (in logarithm of the minimal angle of resolution [logMAR] units) and 25-item National Eye Institute Visual Function Questionnaire composite score change were compared among the 3 groups. Best-corrected visual acuity improved from 1.61 to 0.86 logMAR at 3 months after fitting CL use. Improvement in BCVA in groups 1, 2, and 3 was 0.95 logMAR, 0.82 logMAR, and 0.37 logMAR, respectively. The mean 25-item National Eye Institute Visual Function Questionnaire composite score for the 11 subscales improved from 37.6 ± 16.0 to 58.4 ± 17.4 (P = .000001). All 11 subscores, except that for driving ability, improved significantly. The general vision subscore improved most in group 3, yet the general health subscore improved most in group 1. No serious adverse events attributable to limbal CL use occurred. The tear-exchangeable limbal CL is safe and effective for the improvement of vision and quality of life in Stevens-Johnson syndrome or toxic epidermal necrolysis patients with severe ocular sequelae.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluoreszein Natriumsalz, BioReagent, suitable for fluorescence
Sigma-Aldrich
Fluoreszein Natriumsalz, used as fluorescent tracer
Sigma-Aldrich
Fluoreszein Natriumsalz, p.a.
Supelco
Fluoreszein Natriumsalz, analytical standard