Merck
  • Hepatic differentiation of human embryonic stem cells as microscaled multilayered colonies leading to enhanced homogeneity and maturation.
Hepatic differentiation of human embryonic stem cells as microscaled multilayered colonies leading to enhanced homogeneity and maturation.
Small (Weinheim an der Bergstrasse, Germany) (2014-07-26)
Rui Yao, Jingyu Wang, Xiaokang Li, Da Jung Jung, Hao Qi, Keh Kooi Kee, Yanan Du
ZUSAMMENFASSUNG
Directed differentiation of human embryonic stem cells (hESCs) towards hepatocyte-like cells on planar tissue culture plates has been extensively investigated with great promise to provide alternative cell sources for drug metabolism/toxicity testing. Recently, hepatic differentiation of hESCs in 3D configuration with better mimicry of embryonic liver development represents incremental efforts to improve the differentiation efficiency and cellular maturation. However, most of the present 3D differentiation configurations involved interruptive operations during the multi-staged differentiation process, which might impose unwanted influence on cellular differentiation. Most of the current researches resulted in generation of hepatocytes with high expression of AFP, which is minimally expressed in primary hepatocytes. Here, off-the-shelf micro-stencil arrays are developed to generate adherent multilayered colonies composed of hESCs-derived cells. Uninterrupted cellular differentiation and proliferation is achieved to recapitulate the continuous and multi-stage liver development. Compared with conventional 2D format, the micro-scaled multilayered colonies with uniform and defined sizes constrained within the microwells are composed of more homogenous and mature hepatocyte-like cells with significantly lowered AFP expression and elevated hepatic functions. The multilayered colonies as novel 3D configuration for hepatic differentiation of hESCs represent a significant step toward efficient generation of functional hepatocytes for regenerative medicine and drug discovery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Harnstoff, analytical standard
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
USP
Harnstoff, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Millipore
Harnstoff -Lösung, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Calcein-AM -Lösung, 4 mM in DMSO, ≥90% (HPLC), solution
Harnstoff, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C