Sigma-Aldrich
Black Phosphorus, Crystal, 99.995%
Sigma-Aldrich
Molybdenum disulfide nanoplatelets
Sigma-Aldrich
Molybdenum disulfide, Crystal, 99.995%
Sigma-Aldrich
Tungsten diselenide, Crystal, 99.995%
Sigma-Aldrich
Molybdenum disulfide, dispersion, 0.1-0.5 mg/mL in H2O
Sigma-Aldrich
Tungsten disulfide nanoplatelets, 50-300 nm, thickness < 3 layers, powder
Sigma-Aldrich
Molybdenum disulfide suspension, avg. no. of layers, < 3
Sigma-Aldrich
Tungsten disulfide suspension
Sigma-Aldrich
Tungsten disulfide suspension
Sigma-Aldrich
Molybdenum(IV) disulfide, lithium intercalated
Sigma-Aldrich
Tungsten (IV) disulfide, lithium intercalated
Sigma-Aldrich
Tungsten (IV) diselenide, lithium intercalated
Sigma-Aldrich
Molybdenum (IV) diselenide, lithium intercalated