Merck
  • Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
The Journal of Physical Chemistry C (2014)
Gunjakar JL, et al.