Merck
  • 2-Deoxy-L-ribose inhibits the invasion of thymidine phosphorylase-overexpressing tumors by suppressing matrix metalloproteinase-9.
2-Deoxy-L-ribose inhibits the invasion of thymidine phosphorylase-overexpressing tumors by suppressing matrix metalloproteinase-9.
International journal of cancer (2006-04-29)
Yuichi Nakajima, Misako Haraguchi, Tatsuhiko Furukawa, Masatatsu Yamamoto, Hayao Nakanishi, Masae Tatematsu, Shin-ichi Akiyama
ZUSAMMENFASSUNG
Thymidine phosphorylase (TP), an enzyme involved in pyrimidine metabolism, is identical with an angiogenic factor, platelet-derived endothelial cell growth factor. 2-Deoxy-D-ribose (D-dRib), the degradation product of thymidine generated by TP activity, has been suggested to be a downstream mediator of TP function. 2-Deoxy-L-ribose (L-dRib), a stereoisomer of D-dRib, inhibited the promotion of angiogenesis, tumor growth and metastasis by TP. In our study, we have shown that nude mice inoculated with TP-overexpressing KB/TP cells had shorter survival times than those injected with control KB/CV cells. KB/TP tumors were also more highly invasive than KB/CV tumors in mice. The expression levels of matrix metalloproteinase (MMP)-9 in KB/TP tumors were significantly higher than those in KB/CV tumors. L-dRib and a TP inhibitior, TPI, extended the survival period of KB/TP tumor-bearing mice. L-dRib also reduced MMP-9 mRNA levels in KB/TP tumors. Furthermore, L-dRib suppressed the mRNA level of MMP-9 in cultured KB/TP cells, and the invasive activity of the cells. L-dRib may be useful for the suppression of invasion of TP-expressing tumor cells.