Merck
  • Hydrogen generation by hydrolysis of MgH2 and enhanced kinetics performance of ammonium chloride introducing
Hydrogen generation by hydrolysis of MgH2 and enhanced kinetics performance of ammonium chloride introducing
International Journal of Hydrogen Energy (2015)
Huang M, et al.