Merck
The mucosae-associated epithelial chemokine (MEC/CCL28) modulates immunity in HIV infection.
PloS one (2007-10-04)
Eleonora Castelletti, Sergio Lo Caputo, Louise Kuhn, Manuela Borelli, Johanna Gajardo, Moses Sinkala, Daria Trabattoni, Chipepo Kankasa, Eleonora Lauri, Alberto Clivio, Luca Piacentini, Dorothy H Bray, Grace M Aldrovandi, Donald M Thea, Francisco Veas, Manuela Nebuloni, Francesco Mazzotta, Mario Clerici
ZUSAMMENFASSUNG
CCL28 (MEC) binds to CCR3 and CCR10 and recruits IgA-secreting plasma cells (IgA-ASC) in the mucosal lamina propria (MLP). Mucosal HIV-specific IgA are detected in HIV-infection and exposure. The CCL28 circuit was analyzed in HIV-infected and-exposed individuals and in HIV-unexposed controls; the effect of CCL28 administration on gastrointestinal MLP IgA-ASC was verified in a mouse model. CCL28 was augmented in breast milk (BM) plasma and saliva of HIV-infected and -exposed individuals; CCR3+ and CCR10+ B lymphocytes were increased in these same individuals. Additionally: 1) CCL28 concentration in BM was associated with longer survival in HIV vertically-infected children; and 2) gastro-intestinal mucosal IgA-ASC were significantly increased in VSV-immunized mice receiving CCL28. CCL28 mediates mucosal immunity in HIV exposure and infection. CCL28-including constructs should be considered in mucosal vaccines to prevent HIV infection of the gastro-intestinal MLP via modulation of IgA-ASC.