Merck
New aminocyclitols as modulators of glucosylceramide metabolism.
Organic & biomolecular chemistry (2005-03-24)
Meritxell Egido-Gabás, Pedro Serrano, Josefina Casas, Amadeu Llebaria, Antonio Delgado
ZUSAMMENFASSUNG
A series of 13 aminocyclitol derivatives belonging to two different families is described. Their configuration is governed by the regio- and stereocontrolled epoxide opening of a suitably protected conduritol-B epoxide. Studies on several glycosyl processing enzymes indicate that some of them are good inhibitors of glucosylceramide hydrolase. A rationale to account for preliminary structure-activity relationships is provided.