Merck
  • THE ORGANIC CHEMISTRY NOTEBOOK SERIES, A DIDACTICAL APPROACH,?(I) A THEORETICAL MECHANISTIC APPROACH TO DIASTEROSELECTIVE SYNTHESIS OF CIS-1, 2-DIALKENYLCYCLOPROPANOLS AND SUBSEQUENT OXY-COPE REARRANGEMENT BY JIN KUN CHA ET AL.
THE ORGANIC CHEMISTRY NOTEBOOK SERIES, A DIDACTICAL APPROACH,?(I) A THEORETICAL MECHANISTIC APPROACH TO DIASTEROSELECTIVE SYNTHESIS OF CIS-1, 2-DIALKENYLCYCLOPROPANOLS AND SUBSEQUENT OXY-COPE REARRANGEMENT BY JIN KUN CHA ET AL.
Revista Boliviana de Quimica (2005)
Bravo J.