- A rare case of glycine encephalopathy unveiled by valproate therapy.
A rare case of glycine encephalopathy unveiled by valproate therapy.
Journal of pediatric neurosciences (2015-07-15)
Velusamy Subramanian, Pramila Kadiyala, Praveen Hariharan, E Neeraj
PMID26167219
ZUSAMMENFASSUNG
Glycine encephalopathy (GE) or nonketotic hyperglycinemia is an autosomal recessive disorder due to a primary defect in glycine cleavage enzyme system. It is characterized by elevated levels of glycine in plasma and cerebrospinal fluid usually presenting with seizures, hypotonia, and developmental delay. In our case, paradoxical increase in seizure frequency on starting sodium valproate led us to diagnose GE.