Merck
Effects of L-leucine in 5q- syndrome and other RPS14-deficient erythroblasts.
Leukemia (2012-03-21)
B H Yip, A Pellagatti, C Vuppusetty, A Giagounidis, U Germing, A A Lamikanra, D J Roberts, M Fernandez-Mercado, E-J McDonald, S Killick, J S Wainscoat, J Boultwood