- GRS defective axonal distribution as a potential contributor to distal spinal muscular atrophy type V pathogenesis in a new model of GRS-associated neuropathy.
GRS defective axonal distribution as a potential contributor to distal spinal muscular atrophy type V pathogenesis in a new model of GRS-associated neuropathy.
Journal of chemical neuroanatomy (2014-09-15)
Ah Jung Seo, Byung Sun Park, Junyang Jung
PMID25218976
ZUSAMMENFASSUNG
Distal spinal muscular atrophy type V (dSMA-V), a hereditary axonal neuropathy, is a glycyl-tRNA synthetase (GRS)-associated neuropathy caused by a mutation in GRS. In this study, using an adenovirus vector system equipped with a neuron-specific promoter, we constructed a new GRS-associated neuropathy mouse model. We found that wild-type GRS (WT) is distributed in peripheral axons, dorsal root ganglion (DRG) cell bodies, central axon terminals and motor neuron cell bodies in the mouse model. In contrast, the L129P mutant GRS was localized in DRG and motor neuron cell bodies. Thus, we propose that the disease-causing L129P mutant is linked to a distribution defect in peripheral nerves in vivo.