Merck
  • Cloning and characterization of a new human UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase, designated pp-GalNAc-T13, that is specifically expressed in neurons and synthesizes GalNAc alpha-serine/threonine antigen.
Cloning and characterization of a new human UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase, designated pp-GalNAc-T13, that is specifically expressed in neurons and synthesizes GalNAc alpha-serine/threonine antigen.
The Journal of biological chemistry (2002-10-31)
Yan Zhang, Hiroko Iwasaki, Han Wang, Takashi Kudo, Timothy B Kalka, Thierry Hennet, Tomomi Kubota, Lamei Cheng, Niro Inaba, Masanori Gotoh, Akira Togayachi, Jianming Guo, Hisashi Hisatomi, Kazuyuki Nakajima, Shoko Nishihara, Mitsuru Nakamura, Jamey D Marth, Hisashi Narimatsu
ZUSAMMENFASSUNG
To date, 10 members of the UDP-N-acetyl-alpha-d-galactosamine:polypeptide N-acetylgalactosaminyltransferase (pp-GalNAc-T) family have been cloned and analyzed in human. In this study, we cloned and analyzed a novel human pp-GalNAc-T from an NT2 cell cDNA library, and we named it pp-GalNAc-T13. In amino acid sequences, pp-GalNAc-T13 was highly homologous, showing 84.3% identity, to pp-GalNAc-T1. Real time PCR analysis revealed pp-GalNAc-T13 to be highly and restrictively expressed in the brain and present at very low or undetectable levels in other tissues, in contrast to the ubiquitous expression of pp-GalNAc-T1. pp-GalNAc-T13 was abundantly expressed in all neuroblastoma cells examined and primary cultured neurons but not in glioblastoma cells and primary cultured astrocytes. pp-GalNAc-T13 exhibited much stronger activity to transfer GalNAc to mucin peptides, such as Muc5Ac and MUC7, than did pp-GalNAc-T1. In addition, pp-GalNAc-T13 differed in substrate specificity to pp-GalNAc-T1. pp-GalNAc-T13 was able to form a triplet Tn epitope, three consecutive GalNAc-Ser/Thr structures, on peptides encoded in syndecan-3, a proteoglycan expressed in neurons. pp-GalNAc-T13-deficient mice have been established in a previous work. Immunohistochemical study showed a remarkable decrease in Tn antigen expression in the cerebellum of the pp-GalNAc-T13 knockout mouse. pp-GalNAc-T13 would be a major enzyme responsible for the synthesis of O-glycan and specifically the Tn antigen epitope in neurons.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-GALNT13 antibody produced in rabbit, affinity isolated antibody