Merck
  • Formation of layered magnesium silicate during the aging of magnesium hydroxide-silica mixtures.
Formation of layered magnesium silicate during the aging of magnesium hydroxide-silica mixtures.
Journal of the American Ceramic Society. American Ceramic Society (1998)
Temuujin J, et al.