Merck
  • Nonsense mutations in FGF8 gene causing different degrees of human gonadotropin-releasing deficiency.
Nonsense mutations in FGF8 gene causing different degrees of human gonadotropin-releasing deficiency.
The Journal of clinical endocrinology and metabolism (2010-05-14)
Ericka B Trarbach, Ana Paula Abreu, Leticia Ferreira Gontijo Silveira, Heraldo Mendes Garmes, Maria Tereza M Baptista, Milena Gurgel Teles, Elaine M F Costa, Moosa Mohammadi, Nelly Pitteloud, Berenice B Mendonca, Ana Claudia Latronico
ZUSAMMENFASSUNG
FGFR1 mutations cause isolated hypogonadotropic hypogonadism (IHH) with or without olfactory abnormalities, Kallmann syndrome, and normosmic IHH respectively. Recently, missense mutations in FGF8, a key ligand for fibroblast growth factor receptor (FGFR) 1 in the ontogenesis of GnRH, were identified in IHH patients, thus establishing FGF8 as a novel locus for human GnRH deficiency. Our objective was to analyze the clinical, hormonal, and molecular findings of two familial IHH patients due to FGF8 gene mutations. The entire coding region of the FGF8 gene was amplified and sequenced in two well-phenotyped IHH probands and their relatives. Two unique heterozygous nonsense mutations in FGF8 (p.R127X and p.R129X) were identified in two unrelated IHH probands, which were absent in 150 control individuals. These two mutations, mapped to the core domain of FGF8, impact all four human FGF8 isoforms, and lead to the deletion of a large portion of the protein, generating nonfunctional FGF8 ligands. The p.R127X mutation was identified in an 18-yr-old Kallmann syndrome female. Her four affected siblings with normosmic IHH or delayed puberty also carried the p.R127X mutation. Additional developmental anomalies, including cleft lip and palate and neurosensorial deafness, were also present in this family. The p.R129X mutation was identified in a 30-yr-old man with familial normosmic IHH and severe GnRH deficiency. We identified the first nonsense mutations in the FGF8 gene in familial IHH with variable degrees of GnRH deficiency and olfactory phenotypes, confirming that loss-of-function mutations in FGF8 cause human GnRH deficiency.