Merck
Imidazolepropionic acid as a urinary metabolite of L-histidine.
Biochemical and biophysical research communications (1962-10-17)
N P SEN, P L McGEER, R M PAUL
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-(1H-Imidazol-4-yl)-propionsäure, ≥98.0% (GC)