- Cloning of a balanced translocation breakpoint in the DiGeorge syndrome critical region and isolation of a novel potential adhesion receptor gene in its vicinity.
Cloning of a balanced translocation breakpoint in the DiGeorge syndrome critical region and isolation of a novel potential adhesion receptor gene in its vicinity.
Human molecular genetics (1995-04-01)
S Demczuk, R Aledo, J Zucman, O Delattre, C Desmaze, L Dauphinot, P Jalbert, G A Rouleau, G Thomas, A Aurias
PMID7633403
ZUSAMMENFASSUNG
Deletions of the 22q11.2 have been associated with a wide range of developmental defects (notably DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome and isolated conotruncal cardiac defects) classified under the acronym CATCH 22. A DiGeorge syndrome patient bearing a balanced translocation whose breakpoint maps within the critical region has been previously described. We report the construction of a cosmid contig spanning the translocation breakpoint and the isolation of a gene mapping 10 kb telomeric to the breakpoint. This gene encodes a novel putative adhesion receptor protein, which could play a role in neural crest cells migration, a process which has been proposed to be altered in DiGeorge syndrome.