Merck
  • High-activity MgO-supported CoMo hydrodesulfurization catalysts prepared by non-aqueous impregnation.
High-activity MgO-supported CoMo hydrodesulfurization catalysts prepared by non-aqueous impregnation.
Applied Catalysis. B, Environmental (2015)
Kaluza L, et al.