Merck
Identification of benzethonium chloride in commercial grapefruit seed extracts.
Journal of agricultural and food chemistry (2001-07-17)
G Takeoka, L Dao, R Y Wong, R Lundin, N Mahoney
ZUSAMMENFASSUNG
Commercial grapefruit seed extracts (GSE) were extracted with chloroform. The solvent was evaporated, and the resulting solid was subsequently analyzed by high-performance liquid chromatography, electrospray ionization mass spectrometry, nuclear magnetic resonance (NMR) spectroscopy, and elemental analysis (by proton-induced X-ray emission [PIXE] analysis). The main constituent was identified as benzethonium chloride, a synthetic antimicrobial agent commonly used in cosmetics and other topical applications. This compound comprised 8.03% (n = 2) of the liquid GSE sample. Higher amounts of benzethonium chloride were found in powder GSE samples.