Merck
  • Identification and characterisation of the gene TWIST NEIGHBOR (TWISTNB) located in the microdeletion syndrome 7p21 region.
Identification and characterisation of the gene TWIST NEIGHBOR (TWISTNB) located in the microdeletion syndrome 7p21 region.
Cytogenetic and genome research (2002-11-20)
C Kosan, J Kunz
ZUSAMMENFASSUNG
We have characterised a 2.4-Mb genomic sequence of a smallest region of overlap (SRO) deleted in the human microdeletion syndrome 7p21 by in silico analysis. Patients harbouring this minimal deletion present in addition to the clinical features of Saethre-Chotzen syndrome (MIM 101400) a distinct learning disability. This genomic region shows a very low gene content. Besides the transcription factor gene TWIST, the Histone Deacetylase 9 (HDAC9), Sorting Nexin 13 (SNX13) and an evolutionarily conserved bHLH transcription factor gene Nephew of Atonal 3 (HNATO3) have been detected previously. Here we describe the localisation and characterisation of the TWIST NEIGHBOR (TWISTNB) gene. Comparison of the predicted proteins of human TWISTNB and mouse Twistnb shows a high degree of conservation. Northern blot analysis of human fetal and adult tissues shows ubiquitous expression in all tissues tested.