Merck
  • Assignment of human transforming growth factor-beta type I and type III receptor genes (TGFBR1 and TGFBR3) to 9q33-q34 and 1p32-p33, respectively.
Assignment of human transforming growth factor-beta type I and type III receptor genes (TGFBR1 and TGFBR3) to 9q33-q34 and 1p32-p33, respectively.
Genomics (1995-07-20)
D W Johnson, M Qumsiyeh, M Benkhalifa, D A Marchuk