Merck
Autoimmune lymphoproliferative syndrome-like disease in patients with LRBA mutation.
Clinical immunology (Orlando, Fla.) (2015-05-02)
Shoshana Revel-Vilk, Ute Fischer, Bärbel Keller, Schafiq Nabhani, Laura Gámez-Díaz, Anne Rensing-Ehl, Michael Gombert, Andrea Hönscheid, Hani Saleh, Avraham Shaag, Arndt Borkhardt, Bodo Grimbacher, Klaus Warnatz, Orly Elpeleg, Polina Stepensky
ZUSAMMENFASSUNG
Mutations in LPS-responsive and beige-like anchor (LRBA) gene were recently described in patients with combined immunodeficiency, enteropathy and autoimmune cytopenia. Here, we extend the clinical and immunological phenotypic spectrum of LRBA associated disorders by reporting on three patients from two unrelated families who presented with splenomegaly and lymphadenopathy, cytopenia, elevated double negative T cells and raised serum Fas ligand levels resembling autoimmune lymphoproliferative syndrome (ALPS) and one asymptomatic patient. Homozygous loss of function mutations in LRBA were identified by whole exome analysis. Similar to ALPS patients, Fas mediated apoptosis was impaired in LRBA deficient patients, while apoptosis in response to stimuli of the intrinsic mitochondria mediated apoptotic pathway was even enhanced. This manuscript illustrates the phenotypic overlap of other primary immunodeficiencies with ALPS-like disorders and strongly underlines the necessity of genetic diagnosis in order to provide early correct diagnosis and subsequent care.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Aphidicolin, ≥98% (HPLC), powder
Sigma-Aldrich
Rapamycin aus Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
6-Phosphonohexansäure, 97%
Sigma-Aldrich
Anti-LRBA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-LRBA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2