Merck
Protein microarray characterization of the S-nitrosoproteome.
Molecular & cellular proteomics : MCP (2013-10-10)
Yun-Il Lee, Daniel Giovinazzo, Ho Chul Kang, Yunjong Lee, Jun Seop Jeong, Paschalis-Thomas Doulias, Zhi Xie, Jianfei Hu, Mehdi Ghasemi, Harry Ischiropoulos, Jiang Qian, Heng Zhu, Seth Blackshaw, Valina L Dawson, Ted M Dawson
ZUSAMMENFASSUNG
Nitric oxide (NO) mediates a substantial part of its physiologic functions via S-nitrosylation, however the cellular substrates for NO-mediated S-nitrosylation are largely unknown. Here we describe the S-nitrosoproteome using a high-density protein microarray chip containing 16,368 unique human proteins. We identified 834 potentially S-nitrosylated human proteins. Using a unique and highly specific labeling and affinity capture of S-nitrosylated proteins, 138 cysteine residues on 131 peptides in 95 proteins were determined, defining critical sites of NO's actions. Of these cysteine residues 113 are novel sites of S-nitrosylation. A consensus sequence motif from these 834 proteins for S-nitrosylation was identified, suggesting that the residues flanking the S-nitrosylated cysteine are likely to be the critical determinant of whether the cysteine is S-nitrosylated. We identify eight ubiquitin E3 ligases, RNF10, RNF11, RNF41, RNF141, RNF181, RNF208, WWP2, and UBE3A, whose activities are modulated by S-nitrosylation, providing a unique regulatory mechanism of the ubiquitin proteasome system. These results define a new and extensive set of proteins that are susceptible to NO regulation via S-nitrosylation. Similar approaches could be used to identify other post-translational modification proteomes.