Merck
  • miR-96 induces cisplatin chemoresistance in non-small cell lung cancer cells by downregulating SAMD9.
miR-96 induces cisplatin chemoresistance in non-small cell lung cancer cells by downregulating SAMD9.
Oncology Letters (2015)
Wu L