Merck
  • Determination of trace gold in ores by butyl acetate extraction and flame atomic absorption spectrometry.
Determination of trace gold in ores by butyl acetate extraction and flame atomic absorption spectrometry.
Metallurgical Analysis (2011)
Shan XG and Lin YF.