Merck
  • Cerium or ruthenium catalyzed oxidation of alcohols to carbonyl compounds by means of sodium bromate.
Cerium or ruthenium catalyzed oxidation of alcohols to carbonyl compounds by means of sodium bromate.
Bulletin of the Chemical Society of Japan (1986)
Kanemoto S, et al.