- Characterization of two novel KRAB-domain-containing zinc finger genes, ZNF460 and ZNF461, on human chromosome 19q13.1-->q13.4.
Characterization of two novel KRAB-domain-containing zinc finger genes, ZNF460 and ZNF461, on human chromosome 19q13.1-->q13.4.
Cytogenetic and genome research (2004-03-09)
J Dai, Y Li, C Ji, F Jin, Z Zheng, X Wang, X Sun, X Xu, S Gu, Y Xie, Y Mao
PMID15004467
ZUSAMMENFASSUNG
This study reports the cloning and characterization of two novel human zinc finger protein cDNAs (ZNF460 and ZNF461) from a fetal brain cDNA library. The ZNF460 cDNA is 3,135 bp in length encoding a 562-amino-acid polypeptide and the ZNF461 cDNA is 2,548 bp encoding a 563-amino-acid protein. Both of the proteins contain a KRAB A+B box and eleven C2H2 type zinc finger motifs. ZNF461 shows high similarity with the rat GIOT-1 gene (GIOT1). The ZNF460 gene mapped to 19q13.4 with 3 exons, and ZNF461 mapped to 19q13.1 with 6 exons. Both of the two genes are ubiquitously expressed in normal human tissues and the abundance of the ZNF460 mRNA is relatively low.