Merck
  • Interferon-induced SCYL2 limits release of HIV-1 by triggering PP2A-mediated dephosphorylation of the viral protein Vpu.
Interferon-induced SCYL2 limits release of HIV-1 by triggering PP2A-mediated dephosphorylation of the viral protein Vpu.
Science signaling (2012-10-11)
Kei Miyakawa, Tatsuya Sawasaki, Satoko Matsunaga, Andrey Tokarev, Gary Quinn, Hirokazu Kimura, Masako Nomaguchi, Akio Adachi, Naoki Yamamoto, John Guatelli, Akihide Ryo
ZUSAMMENFASSUNG
Human cells respond to infection by retroviruses through the actions of proteins that inhibit the spread of viruses to other cells. One example is bone marrow stromal cell antigen 2 (BST2; also known as tetherin), which is an interferon (IFN)-inducible protein that restricts the release of progeny virions from infected cells. The HIV-1 accessory protein Vpu (viral protein U) causes degradation of BST2, and phosphorylation of Vpu at residues Ser(52) and Ser(56) is required for this function. We report that the host protein SCY1-like protein 2 (SCYL2) mediates the dephosphorylation of Vpu, antagonizing Vpu function and facilitating BST2-dependent restriction of HIV-1 release. SCYL2 reduced the number of virus particles released from cells infected with wild-type HIV-1, but not a strain lacking vpu, in a BST2-dependent manner. SCYL2 stimulated the dephosphorylation of Vpu on Ser(52) and Ser(56) by recruiting protein phosphatase 2A (PP2A) to Vpu. Conversely, depletion of SCYL2 resulted in enhanced phosphorylation of Vpu and increased viral particle release. Moreover, SCYL2 was produced in response to type I IFN and contributed to IFN-mediated viral restriction. Together, these results suggest that SCYL2 serves as a regulatory factor for Vpu, reducing the extent of Vpu phosphorylation and consequently enhancing BST2-mediated viral restriction.