Merck
  • Chromosome aberrations involving 10q22: report of three overlapping interstitial deletions and a balanced translocation disrupting C10orf11.
Chromosome aberrations involving 10q22: report of three overlapping interstitial deletions and a balanced translocation disrupting C10orf11.
European journal of human genetics : EJHG (2009-10-22)
Andreas Tzschach, Anne-Marie Bisgaard, Maria Kirchhoff, Luitgard M Graul-Neumann, Heidemarie Neitzel, Stephanie Page, Alischo Ahmed, Ines Müller, Fikret Erdogan, Hans-Hilger Ropers, Vera M Kalscheuer, Reinhard Ullmann
ZUSAMMENFASSUNG
Interstitial deletions of chromosome band 10q22 are rare. We report on the characterization of three overlapping de novo 10q22 deletions by high-resolution array comparative genomic hybridization in three unrelated patients. Patient 1 had a 7.9 Mb deletion in 10q21.3-q22.2 and suffered from severe feeding problems, facial dysmorphisms and profound mental retardation. Patients 2 and 3 had nearly identical deletions of 3.2 and 3.6 Mb, the proximal breakpoints of which were located at an identical low-copy repeat. Both patients were mentally retarded; patient 3 also suffered from growth retardation and hypotonia. We also report on the results of breakpoint analysis by array painting in a mentally retarded patient with a balanced chromosome translocation 46,XY,t(10;13)(q22;p13)dn. The breakpoint in 10q22 was found to disrupt C10orf11, a brain-expressed gene in the common deleted interval of patients 1-3. This finding suggests that haploinsufficiency of C10orf11 contributes to the cognitive defects in 10q22 deletion patients.