Merck
  • Silencing of Peroxiredoxin 2 and aberrant methylation of 33 CpG islands in putative promoter regions in human malignant melanomas.
Silencing of Peroxiredoxin 2 and aberrant methylation of 33 CpG islands in putative promoter regions in human malignant melanomas.
Cancer research (2006-06-17)
Junichi Furuta, Yoshimasa Nobeyama, Yoshihiro Umebayashi, Fujio Otsuka, Kanako Kikuchi, Toshikazu Ushijima
ZUSAMMENFASSUNG
Aberrant methylation of promoter CpG islands (CGI) is involved in silencing of tumor suppressor genes and is also a potential cancer biomarker. Here, to identify CGIs aberrantly methylated in human melanomas, we did a genome-wide search using methylation-sensitive representational difference analysis. CGIs in putative promoter regions of 34 genes (ABHD9, BARHL1, CLIC5, CNNM1, COL2A1, CPT1C, DDIT4L, DERL3, DHRS3, DPYS, EFEMP2, FAM62C, FAM78A, FLJ33790, GBX2, GPR10, GPRASP1, HOXA9, HOXD11, HOXD12, HOXD13, p14ARF, PAX6, PRDX2, PTPRG, RASD1, RAX, REC8L1, SLC27A3, TGFB2, TLX2, TMEM22, TMEM30B, and UNC5C) were found to be methylated in at least 1 of 13 melanoma cell lines but not in two cultured normal melanocytes. Among these genes, Peroxiredoxin 2 (PRDX2) was expressed in normal melanocytes, and its expression was lost in melanomas with methylation. The loss of expression was restored by treatment of melanomas with a demethylating agent 5-aza-2'-deoxycytidine. In surgical melanoma specimens, methylation of PRDX2 was detected in 3 of 36 (8%). Furthermore, immunohistochemical analysis of PRDX2 showed that disappearance of immunoreactivity tends to associate with its methylation. PRDX2 was recently reported to be a negative regulator of platelet-derived growth factor signaling, and its silencing was suggested to be involved in melanomas. On the other hand, 12 CGIs were methylated in >or=9 of the 13 melanoma cell lines and are considered as candidate melanoma biomarkers.