Merck
  • Low-temperature atomic layer deposition of Al2O3 thin coatings for corrosion protection of steel: surface and electrochemical analysis.
Low-temperature atomic layer deposition of Al<SUB>2</SUB>O<SUB>3</SUB> thin coatings for corrosion protection of steel: surface and electrochemical analysis.
Corrosion Science (2011)
Diaz B, et al.