Merck
  • Enhancing antimicrobial activity of chitosan films by incorporating garlic oil, potassium sorbate and nisin.
Enhancing antimicrobial activity of chitosan films by incorporating garlic oil, potassium sorbate and nisin.
LWT--Food Science and Technology (2005)
Pranoto Y, et al.