Merck
  • Construction of 3, 5-substituted 1, 2, 4-oxadiazole rings triggered by tetrabutylammonium hydroxide: a highly efficient and fluoride-free ring closure reaction of O-acylamidoximes.
Construction of 3, 5-substituted 1, 2, 4-oxadiazole rings triggered by tetrabutylammonium hydroxide: a highly efficient and fluoride-free ring closure reaction of <I>O</I>-acylamidoximes.
Tetrahedron Letters (2014)
Otaka H, et al.