Merck
  • Structural and DC electrical investigations of ZnO thin films prepared by spray pyrolysis technique.
Structural and DC electrical investigations of ZnO thin films prepared by spray pyrolysis technique.
Physica B: Condensed Matter (Amsterdam, Netherlands) (2001)
Riad AS, et al.